Assay Details
| Gene Symbol: AT3G13070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112146.3 | NP_187914.1 | 9 - 10 | 1666 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G13070| Gene Symbol: AT3G13070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112146.3 | NP_187914.1 | 9 - 10 | 1666 | 94 | |
Set Membership: |
|