Assay Details
| Gene Symbol: ATSPO11-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112156.2 | NP_187923.1 | 12 - 13 | 789 | 104 | |
| GenBank mRNA | AJ251989.1 | - | 12 - 13 | 828 | 104 | |
| BT028955.1 | - | 12 - 13 | 775 | 104 | ||
More Information
Set Membership: |
|

ATSPO11-1
Genomic Map