Assay Details
| Gene Symbol: ATSPO11-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338034.1 | NP_001326966.1 | 8 - 9 | 535 | 71 | |
| NM_001338035.1 | NP_001326967.1 | 8 - 9 | 521 | 71 | ||
| NM_112156.2 | NP_187923.1 | 8 - 9 | 535 | 71 | ||
| GenBank mRNA | AJ251989.1 | - | 8 - 9 | 574 | 71 | |
| BT028955.1 | - | 8 - 9 | 521 | 71 | ||
More Information
Set Membership: |
|

ATSPO11-1
Genomic Map