Assay Details
| Gene Symbol: AT3G13180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112158.5 | NP_187924.2 | 13 - 14 | 1334 | 110 | |
| GenBank mRNA | AK226254.1 | - | 13 - 14 | 1197 | 110 | |
| AY072189.1 | - | 13 - 14 | 1227 | 110 | ||
| AY117322.1 | - | 13 - 14 | 1184 | 110 | ||
More Information
Set Membership: |
|

AT3G13180
Genomic Map