Assay Details
| Gene Symbol: AT3G13180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112158.5 | NP_187924.2 | 16 - 17 | 1586 | 69 | |
| GenBank mRNA | AY072189.1 | - | 16 - 17 | 1479 | 69 | |
| AY117322.1 | - | 16 - 17 | 1436 | 69 | ||
More Information
Set Membership: |
|

AT3G13180
Genomic Map