Assay Details
| Gene Symbol: AT3G13180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112158.5 | NP_187924.2 | 7 - 8 | 835 | 75 | |
| GenBank mRNA | AK226254.1 | - | 7 - 8 | 698 | 75 | |
| AY072189.1 | - | 7 - 8 | 728 | 75 | ||
| AY117322.1 | - | 7 - 8 | 685 | 75 | ||
More Information
Set Membership: |
|

AT3G13180
Genomic Map