Assay Details
| Gene Symbol: AT3G13180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112158.5 | NP_187924.2 | 9 - 10 | 988 | 71 | |
| GenBank mRNA | AK226254.1 | - | 9 - 10 | 851 | 71 | |
| AY072189.1 | - | 9 - 10 | 881 | 71 | ||
| AY117322.1 | - | 9 - 10 | 838 | 71 | ||
More Information
Set Membership: |
|

AT3G13180
Genomic Map