Assay Details
| Gene Symbol: TBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035611.2 | NP_001030688.1 | 4 - 5 | 500 | 79 | |
| NM_112190.5 | NP_187953.1 | 4 - 5 | 503 | 79 | ||
| GenBank mRNA | AB493611.1 | - | 3 - 4 | 238 | 79 | |
| AF324696.3 | - | 4 - 5 | 502 | 79 | ||
| AF327429.1 | - | 4 - 5 | 498 | 79 | ||
| AF349523.1 | - | 3 - 4 | 238 | 79 | ||
| AY054608.1 | - | 4 - 5 | 498 | 79 | ||
| AY072455.1 | - | 3 - 4 | 238 | 79 | ||
| AY084881.1 | - | 4 - 5 | 488 | 79 | ||
| AY463625.1 | - | 3 - 4 | 238 | 79 | ||
| X54996.1 | - | 465 | 79 | |||
More Information
Set Membership: |
|

TBP1
Genomic Map