Assay Details
| Gene Symbol: TBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112190.5 | NP_187953.1 | 8 - 9 | 787 | 115 | |
| GenBank mRNA | AB493611.1 | - | 7 - 8 | 522 | 115 | |
| AF324696.3 | - | 8 - 9 | 786 | 115 | ||
| AF327429.1 | - | 8 - 9 | 782 | 115 | ||
| AF349523.1 | - | 7 - 8 | 522 | 115 | ||
| AY054608.1 | - | 8 - 9 | 782 | 115 | ||
| AY072455.1 | - | 7 - 8 | 522 | 115 | ||
| AY084881.1 | - | 8 - 9 | 772 | 115 | ||
| AY463625.1 | - | 7 - 8 | 522 | 115 | ||
| X54996.1 | - | 749 | 115 | |||
More Information
Set Membership: |
|

TBP1
Genomic Map