Assay Details
| Gene Symbol: PUT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112208.5 | NP_566460.1 | 1 - 2 | 364 | 79 | |
| GenBank mRNA | AY056273.1 | - | 1 - 2 | 298 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PUT4| Gene Symbol: PUT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112208.5 | NP_566460.1 | 1 - 2 | 364 | 79 | |
| GenBank mRNA | AY056273.1 | - | 1 - 2 | 298 | 79 | |
Set Membership: |
|