Assay Details
| Gene Symbol: PRA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112222.4 | NP_566461.1 | 1 | 1307 | 94 | |
| GenBank mRNA | BT004174.1 | - | 1 | 325 | 94 | |
| BT004952.1 | - | 1 | 275 | 94 | ||
More Information
Set Membership: |
|

PRA8
Genomic Map