Assay Details
| Gene Symbol: BGAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112225.5 | NP_187988.1 | 10 - 11 | 1324 | 78 | |
| GenBank mRNA | AJ270297.1 | - | 10 - 11 | 1302 | 78 | |
More Information
Set Membership: |
|

BGAL1
Genomic Map