Assay Details
| Gene Symbol: RHD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112241.4 | NP_188003.1 | 21 - 22 | 2337 | 102 | |
| NM_202579.1 | NP_974308.1 | 21 - 22 | 2246 | 102 | ||
| GenBank mRNA | AY092988.1 | - | 21 - 22 | 2181 | 102 | |
| AY128801.1 | - | 20 - 21 | 2091 | 102 | ||
More Information
Set Membership: |
|

RHD3
Genomic Map