Assay Details
| Gene Symbol: AT3G13930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112247.3 | NP_566470.1 | 9 - 10 | 847 | 132 | |
| GenBank mRNA | AF367302.1 | - | 9 - 10 | 677 | 132 | |
| AY091691.1 | - | 8 - 9 | 537 | 132 | ||
| BT000444.1 | - | 9 - 10 | 677 | 132 | ||
| BT000702.1 | - | 9 - 10 | 679 | 132 | ||
| BT001223.1 | - | 8 - 9 | 537 | 132 | ||
More Information
Set Membership: |
|

AT3G13930
Genomic Map