Assay Details
| Gene Symbol: CID4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084681.1 | NP_001078150.1 | 14 - 15 | 2021 | 108 | |
| NM_001084682.1 | NP_001078151.1 | 14 - 15 | 1998 | 108 | ||
| NM_001338097.1 | NP_001325470.1 | 14 - 15 | 2545 | 108 | ||
| NM_112255.4 | NP_566471.1 | 14 - 15 | 2329 | 108 | ||
| GenBank mRNA | AY045922.1 | - | 14 - 15 | 2025 | 108 | |
| AY079365.1 | - | 13 - 14 | 1759 | 108 | ||
More Information
Set Membership: |
|

CID4
Genomic Map