Assay Details
| Gene Symbol: CID4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084681.1 | NP_001078150.1 | 4 - 5 | 650 | 67 | |
| NM_001084682.1 | NP_001078151.1 | 4 - 5 | 627 | 67 | ||
| NM_001202957.1 | NP_001189886.1 | 4 - 5 | 767 | 67 | ||
| NM_001338097.1 | NP_001325470.1 | 4 - 5 | 1174 | 67 | ||
| NM_112255.4 | NP_566471.1 | 4 - 5 | 958 | 67 | ||
| GenBank mRNA | AY045922.1 | - | 4 - 5 | 654 | 67 | |
| AY079365.1 | - | 3 - 4 | 388 | 67 | ||
More Information
Set Membership: |
|

CID4
Genomic Map