Assay Details
| Gene Symbol: CID4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084681.1 | NP_001078150.1 | 7 - 8 | 1236 | 90 | |
| NM_001084682.1 | NP_001078151.1 | 7 - 8 | 1213 | 90 | ||
| NM_001338097.1 | NP_001325470.1 | 7 - 8 | 1760 | 90 | ||
| NM_112255.4 | NP_566471.1 | 7 - 8 | 1544 | 90 | ||
| GenBank mRNA | AY045922.1 | - | 7 - 8 | 1240 | 90 | |
| AY079365.1 | - | 6 - 7 | 974 | 90 | ||
More Information
Set Membership: |
|

CID4
Genomic Map