Assay Details
| Gene Symbol: RSH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112259.5 | NP_188021.1 | 1 - 2 | 1212 | 125 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RSH2| Gene Symbol: RSH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112259.5 | NP_188021.1 | 1 - 2 | 1212 | 125 | |
Set Membership: |
|