Assay Details
| Gene Symbol: AT3G14067 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112261.4 | NP_566473.2 | 1 | 2485 | 58 | |
| GenBank mRNA | AK229057.1 | - | 1 | 2321 | 58 | |
| BT011692.1 | - | 1 | 2322 | 58 | ||
| BT012275.1 | - | 1 | 2294 | 58 | ||
More Information
Set Membership: |
|

AT3G14067
Genomic Map