Assay Details
| Gene Symbol: AT3G14200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112276.4 | NP_188036.1 | 1 - 2 | 444 | 66 | |
| GenBank mRNA | AK227818.1 | - | 1 - 2 | 344 | 66 | |
| AY091107.1 | - | 1 - 2 | 398 | 66 | ||
| AY122970.1 | - | 1 - 2 | 107 | 66 | ||
More Information
Set Membership: |
|

AT3G14200
Genomic Map