Assay Details
| Gene Symbol: SRF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112293.5 | NP_188052.2 | 10 - 11 | 1276 | 109 | |
| NM_202582.1 | NP_974311.1 | 9 - 10 | 1332 | 109 | ||
| NM_202583.1 | NP_974312.1 | 10 - 11 | 1200 | 109 | ||
| GenBank mRNA | AY518292.1 | - | 10 - 11 | 1270 | 109 | |
| BT046202.1 | - | 10 - 11 | 1032 | 109 | ||
| FJ708721.1 | - | 10 - 11 | 1032 | 109 | ||
More Information
Set Membership: |
|

SRF7
Genomic Map