Assay Details
| Gene Symbol: SRF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112293.5 | NP_188052.2 | 14 - 15 | 2108 | 78 | |
| NM_202582.1 | NP_974311.1 | 13 - 14 | 2164 | 78 | ||
| NM_202583.1 | NP_974312.1 | 15 - 16 | 1948 | 78 | ||
| GenBank mRNA | AY518292.1 | - | 14 - 15 | 2102 | 78 | |
| BT046202.1 | - | 14 - 15 | 1864 | 78 | ||
| FJ708721.1 | - | 14 - 15 | 1864 | 78 | ||
More Information
Set Membership: |
|

SRF7
Genomic Map