Assay Details
| Gene Symbol: SRF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112293.5 | NP_188052.2 | 3 - 4 | 545 | 84 | |
| NM_202582.1 | NP_974311.1 | 2 - 3 | 601 | 84 | ||
| NM_202583.1 | NP_974312.1 | 3 - 4 | 469 | 84 | ||
| GenBank mRNA | AY518292.1 | - | 3 - 4 | 539 | 84 | |
| BT046202.1 | - | 3 - 4 | 301 | 84 | ||
| FJ708721.1 | - | 3 - 4 | 301 | 84 | ||
More Information
Set Membership: |
|

SRF7
Genomic Map