Assay Details
| Gene Symbol: CUC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112380.3 | NP_188135.1 | 1 - 2 | 326 | 115 | |
| GenBank mRNA | AB049069.1 | - | 1 - 2 | 212 | 115 | |
| BT026080.1 | - | 1 - 2 | 196 | 115 | ||
More Information
Set Membership: |
|

CUC1
Genomic Map