Assay Details
| Gene Symbol: CUC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112380.3 | NP_188135.1 | 2 - 3 | 613 | 83 | |
| GenBank mRNA | AB049069.1 | - | 2 - 3 | 499 | 83 | |
| BT026080.1 | - | 2 - 3 | 483 | 83 | ||
More Information
Set Membership: |
|

CUC1
Genomic Map