Assay Details
| Gene Symbol: REM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342099.1 | NP_001328023.1 | 5 - 6 | 1371 | 99 | |
| NM_119310.4 | NP_567880.1 | 5 - 6 | 1356 | 99 | ||
| GenBank mRNA | BT003891.1 | - | 5 - 6 | 1356 | 99 | |
| BT005002.1 | - | 5 - 6 | 1155 | 99 | ||
More Information
Set Membership: |
|

REM1
Genomic Map