Assay Details
| Gene Symbol: AT4G31810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119331.6 | NP_194909.2 | 5 - 6 | 565 | 92 | |
| GenBank mRNA | AY080787.1 | - | 5 - 6 | 444 | 92 | |
| AY114019.1 | - | 5 - 6 | 425 | 92 | ||
More Information
Set Membership: |
|

AT4G31810
Genomic Map