Assay Details
| Gene Symbol: AT4G31875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119338.2 | NP_567884.1 | 1 | 707 | 116 | |
| GenBank mRNA | AY057635.1 | - | 1 | 545 | 116 | |
| Gene Symbol: GPX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342119.1 | NP_001328811.1 | 6 | 971 | 116 | |
| NM_119337.3 | NP_194915.2 | 6 | 957 | 116 | ||
More Information
Set Membership: |
|

AT4G31875
Genomic Map