Assay Details
| Gene Symbol: HSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342124.1 | NP_001329752.1 | 8 - 9 | 1553 | 72 | |
| NM_119353.4 | NP_194929.2 | 10 - 11 | 1913 | 72 | ||
| GenBank mRNA | AB206554.1 | - | 6 - 7 | 1099 | 72 | |
| AK228013.1 | - | 10 - 11 | 1756 | 72 | ||
More Information
Set Membership: |
|

HSL1
Genomic Map