Assay Details
| Gene Symbol: HSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342124.1 | NP_001329752.1 | 9 - 10 | 1633 | 69 | |
| NM_119353.4 | NP_194929.2 | 11 - 12 | 1993 | 69 | ||
| GenBank mRNA | AB206554.1 | - | 7 - 8 | 1179 | 69 | |
| AK228013.1 | - | 11 - 12 | 1836 | 69 | ||
More Information
Set Membership: |
|

HSL1
Genomic Map