Assay Details
| Gene Symbol: HSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342124.1 | NP_001329752.1 | 12 - 13 | 2117 | 90 | |
| NM_119353.4 | NP_194929.2 | 14 - 15 | 2477 | 90 | ||
| GenBank mRNA | AB206554.1 | - | 10 - 11 | 1663 | 90 | |
| AK228013.1 | - | 14 - 15 | 2320 | 90 | ||
More Information
Set Membership: |
|

HSL1
Genomic Map