Assay Details
| Gene Symbol: HSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119353.4 | NP_194929.2 | 8 - 9 | 1646 | 84 | |
| GenBank mRNA | AB206554.1 | - | 4 - 5 | 832 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HSL1| Gene Symbol: HSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119353.4 | NP_194929.2 | 8 - 9 | 1646 | 84 | |
| GenBank mRNA | AB206554.1 | - | 4 - 5 | 832 | 84 | |
Set Membership: |
|