Assay Details
| Gene Symbol: PANK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036691.1 | NP_001031768.1 | 10 - 11 | 1585 | 124 | |
| NM_001125620.1 | NP_001119092.1 | 10 - 11 | 1585 | 124 | ||
| NM_119370.5 | NP_194945.3 | 10 - 11 | 1635 | 124 | ||
More Information
Set Membership: |
|

PANK2
Genomic Map