Assay Details
| Gene Symbol: La1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036700.2 | NP_001031777.1 | 8 - 9 | 1241 | 88 | |
| NM_119425.5 | NP_567904.1 | 8 - 9 | 1256 | 88 | ||
| GenBank mRNA | AY056237.1 | - | 8 - 9 | 1180 | 88 | |
More Information
Set Membership: |
|

La1
Genomic Map