Assay Details
| Gene Symbol: La1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119425.5 | NP_567904.1 | 9 - 10 | 1332 | 86 | |
| GenBank mRNA | AY056237.1 | - | 9 - 10 | 1256 | 86 | |
More Information
Set Membership: |
|

La1
Genomic Map