Assay Details
| Gene Symbol: PC-MYB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342169.1 | NP_001328944.1 | 5 - 6 | 812 | 116 | |
| NM_001342170.1 | NP_001328945.1 | 5 - 6 | 600 | 116 | ||
| NM_001342171.1 | NP_001328946.1 | 5 - 6 | 585 | 116 | ||
| NM_119426.2 | NP_194999.1 | 5 - 6 | 585 | 116 | ||
| NM_202989.3 | NP_974718.1 | 5 - 6 | 797 | 116 | ||
| GenBank mRNA | AF151646.1 | - | 4 - 5 | 277 | 116 | |
| AF176005.2 | - | 5 - 6 | 585 | 116 | ||
More Information
Set Membership: |
|

PC-MYB1
Genomic Map