Assay Details
| Gene Symbol: VTE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119430.5 | NP_567906.1 | 8 - 9 | 1331 | 115 | |
| GenBank mRNA | AF302188.1 | - | 8 - 9 | 1134 | 115 | |
| AK226552.1 | - | 8 - 9 | 1201 | 115 | ||
More Information
Set Membership: |
|

VTE1
Genomic Map