Assay Details
| Gene Symbol: GLDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119455.3 | NP_195027.1 | 12 - 13 | 3175 | 74 | |
| GenBank mRNA | AY063903.1 | - | 12 - 13 | 3053 | 74 | |
| AY091186.1 | - | 12 - 13 | 2903 | 74 | ||
More Information
Set Membership: |
|

GLDP1
Genomic Map