Assay Details
| Gene Symbol: GLDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125634.1 | NP_001119106.1 | 2 - 3 | 1647 | 91 | |
| NM_119455.3 | NP_195027.1 | 2 - 3 | 1737 | 91 | ||
| GenBank mRNA | AY063903.1 | - | 2 - 3 | 1615 | 91 | |
| AY091186.1 | - | 2 - 3 | 1465 | 91 | ||
More Information
Set Membership: |
|

GLDP1
Genomic Map