Assay Details
| Gene Symbol: AT4G33060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119460.5 | NP_195032.2 | 4 - 5 | 683 | 80 | |
| GenBank mRNA | AY568525.1 | - | 4 - 5 | 603 | 80 | |
| BT025329.1 | - | 4 - 5 | 603 | 80 | ||
More Information
Set Membership: |
|

AT4G33060
Genomic Map