Assay Details
Gene Symbol: ADR1-L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036704.2 | NP_001031781.1 | 3 - 4 | 1446 | 97 | |
NM_001342211.1 | NP_001320123.1 | 2 - 3 | 2145 | 97 | ||
GenBank mRNA | AK316970.1 | - | 2 - 3 | 1078 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
