Assay Details
| Gene Symbol: HDA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119501.4 | NP_567921.1 | 7 - 8 | 901 | 115 | |
| GenBank mRNA | AY052234.1 | - | 7 - 8 | 855 | 115 | |
| AY113069.1 | - | 7 - 8 | 790 | 115 | ||
More Information
Set Membership: |
|

HDA14
Genomic Map