Assay Details
| Gene Symbol: PAA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342218.1 | NP_001328919.1 | 1 - 2 | 540 | 81 | |
| NM_119506.4 | NP_567924.1 | 1 - 2 | 602 | 81 | ||
| NM_202946.3 | NP_974675.1 | 1 - 2 | 627 | 81 | ||
| NM_202947.1 | NP_974676.1 | 1 - 2 | 561 | 81 | ||
| GenBank mRNA | AK229051.1 | - | 1 - 2 | 530 | 81 | |
| AY059869.1 | - | 1 - 2 | 541 | 81 | ||
| AY093320.1 | - | 1 - 2 | 464 | 81 | ||
More Information
Set Membership: |
|

PAA1
Genomic Map