Assay Details
| Gene Symbol: PAA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119506.4 | NP_567924.1 | 3 - 4 | 810 | 156 | |
| GenBank mRNA | AY059869.1 | - | 3 - 4 | 749 | 156 | |
More Information
Set Membership: |
|

PAA1
Genomic Map