Assay Details
| Gene Symbol: PAA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342218.1 | NP_001328919.1 | 4 - 5 | 1114 | 100 | |
| NM_119506.4 | NP_567924.1 | 4 - 5 | 897 | 100 | ||
| GenBank mRNA | AY059869.1 | - | 4 - 5 | 836 | 100 | |
More Information
Set Membership: |
|

PAA1
Genomic Map