Assay Details
| Gene Symbol: EX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119519.4 | NP_567929.1 | 11 - 12 | 2378 | 101 | |
| GenBank mRNA | AY059742.1 | - | 11 - 12 | 2167 | 101 | |
More Information
Set Membership: |
|

EX1
Genomic Map