Assay Details
| Gene Symbol: AGD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119526.4 | NP_567934.1 | 7 - 8 | 663 | 69 | |
| GenBank mRNA | AY056423.1 | - | 7 - 8 | 621 | 69 | |
| AY065256.1 | - | 7 - 8 | 615 | 69 | ||
| AY117246.1 | - | 7 - 8 | 570 | 69 | ||
| AY518701.1 | - | 7 - 8 | 570 | 69 | ||
More Information
Set Membership: |
|

AGD2
Genomic Map