Assay Details
| Gene Symbol: AGD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119526.4 | NP_567934.1 | 8 - 9 | 979 | 71 | |
| GenBank mRNA | AY056423.1 | - | 8 - 9 | 937 | 71 | |
| AY065256.1 | - | 8 - 9 | 931 | 71 | ||
| AY117246.1 | - | 8 - 9 | 886 | 71 | ||
| AY518701.1 | - | 8 - 9 | 886 | 71 | ||
More Information
Set Membership: |
|

AGD2
Genomic Map