Assay Details
| Gene Symbol: AT4G33945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119555.4 | NP_567944.1 | 3 - 4 | 966 | 68 | |
| GenBank mRNA | AY099835.1 | - | 3 - 4 | 800 | 68 | |
| BT026356.1 | - | 3 - 4 | 731 | 68 | ||
More Information
Set Membership: |
|

AT4G33945
Genomic Map