Assay Details
| Gene Symbol: AT4G33945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119555.4 | NP_567944.1 | 7 - 8 | 1497 | 93 | |
| GenBank mRNA | AY099835.1 | - | 7 - 8 | 1331 | 93 | |
| BT026356.1 | - | 7 - 8 | 1262 | 93 | ||
More Information
Set Membership: |
|

AT4G33945
Genomic Map